4-Hydroxy-3-methoxybenzotrifluoride

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methoxy-4-(trifluoromethyl)phenol

PC48583-1G PC48583-250MG
APOSPC48583-1GEA 207 EUR
APOSPC48583-1G APOSPC48583-250MG
4-Hydroxy-3-methoxybenzotrifluoride
4-Hydroxy-3-methoxybenzotrifluoride
Formel: C₈H₇F₃O₂
Molecular Weight: 192,135 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13185693
CAS-Nummer: 1027888-79-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR