Methyl 4-bromo-3-fluoro-2-hydroxybenzoate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 4-bromo-3-fluorosalicylate, 3-Bromo-2-fluoro-6-(methoxycarbonyl)phenol
PC48605-1G
PC48605-250MG
PC48605-100MG
PC48605-5G
PC48605-25G
APOSPC48605-1GEA
328
EUR
APOSPC48605-1G
APOSPC48605-250MG
APOSPC48605-100MG
APOSPC48605-5G
APOSPC48605-25G
Methyl 4-bromo-3-fluoro-2-hydroxybenzoate
Methyl 4-bromo-3-fluoro-2-hydroxybenzoate
Formel:
C₈H₆BrFO₃ Molecular Weight: 249,034 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD28122826 CAS-Nummer: 1807191-62-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...