4,4,5,5-Tetramethyl-2-(2,3,4-trifluorophenyl)-1,3,2-dioxaborolane

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3,4-Trifluorophenylboronic acid pinacol ester

PC48625-1G PC48625-250MG
APOSPC48625-1GEA 140 EUR
APOSPC48625-1G APOSPC48625-250MG
4,4,5,5-Tetramethyl-2-(2,3,4-trifluorophenyl)-1,3,2-dioxaborolane
4,4,5,5-Tetramethyl-2-(2,3,4-trifluorophenyl)-1,3,2-dioxaborolane
Formel: C₁₂H₁₄BF₃O₂
Molecular Weight: 258,045 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12405349
CAS-Nummer: 881401-96-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR