2-(5-Bromo-2,3,4-trifluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC48626-1G
PC48626-250MG
APOSPC48626-1GEA
220
EUR
APOSPC48626-1G
APOSPC48626-250MG
2-(5-Bromo-2,3,4-trifluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(5-Bromo-2,3,4-trifluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formel:
C₁₂H₁₃BBrF₃O₂ Molecular Weight: 336,941 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12405350 CAS-Nummer: 1073339-18-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...