2-(5-Bromo-2-fluoro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Lieferant: Apollo Scientific
Achtung

PC48628-1G PC48628-250MG
APOSPC48628-1GEA 114 EUR
APOSPC48628-1G APOSPC48628-250MG
2-(5-Bromo-2-fluoro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2-(5-Bromo-2-fluoro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formel: C₁₃H₁₇BBrFO₂
Molecular Weight: 314,986 g/mol
Schmelzpunkt: 70…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12405354
CAS-Nummer: 1111096-03-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR