5-Bromo-2,3,4-trifluorophenol 95%

Lieferant: Apollo Scientific
Achtung

PC48631-1G PC48631-250MG PC48631-5G
APOSPC48631-1GEA 284 EUR
APOSPC48631-1G APOSPC48631-250MG APOSPC48631-5G
5-Bromo-2,3,4-trifluorophenol 95%
5-Bromo-2,3,4-trifluorophenol
Formel: C₆H₂BrF₃O
Molecular Weight: 226,979 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12405433
CAS-Nummer: 1073339-19-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR