3-Methoxy-5-methylbenzotrifluoride 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-methyl-5-(trifluoromethyl)phenyl ether, 1-Methoxy-3-methyl-5-(trifluoromethyl)benzene

PC48636-1G PC48636-250MG
APOSPC48636-1GEA 551 EUR
APOSPC48636-1G APOSPC48636-250MG
3-Methoxy-5-methylbenzotrifluoride 95%
3-Methoxy-5-methylbenzotrifluoride
Formel: C₉H₉F₃O
Molecular Weight: 190,162 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185712
CAS-Nummer: 1214385-12-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR