2,6-Difluoro-3-(methoxymethoxy)benzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC48637-1G
PC48637-500MG
APOSPC48637-1GEA
1570
EUR
APOSPC48637-1G
APOSPC48637-500MG
2,6-Difluoro-3-(methoxymethoxy)benzoic acid
2,6-Difluoro-3-(methoxymethoxy)benzoic acid
Formel:
C₉H₈F₂O₄ Molecular Weight: 218.154 g/mol Schmelzpunkt: 70…73 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD22689854 CAS-Nummer: 1190603-51-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...