4'-Bromo-2,2,2,2',6'-pentafluoroacetophenone
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(4-Bromo-2,6-difluorophenyl)-2,2,2-trifluoroethanone
PC48641-1G
PC48641-250MG
PC48641-5G
APOSPC48641-1GEA
212
EUR
APOSPC48641-1G
APOSPC48641-250MG
APOSPC48641-5G
4'-Bromo-2,2,2,2',6'-pentafluoroacetophenone
4'-Bromo-2,2,2,2',6'-pentafluoroacetophenone
Formel:
C₈H₂BrF₅O Molecular Weight: 288,997 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12922633 CAS-Nummer: 1208074-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...