6-Bromo-3-chloro-2,4-difluorobenzaldehyde 98%
Lieferant: Apollo Scientific
PC48651-1G
APOSPC48651-1GEA
377
EUR
APOSPC48651-1G
6-Bromo-3-chloro-2,4-difluorobenzaldehyde 98%
6-Bromo-3-chloro-2,4-difluorobenzaldehyde
Formel:
C₇H₂BrClF₂O Molecular Weight: 255,444 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11845975 CAS-Nummer: 1160573-22-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...