4-(Trifluoromethyl)pyridine-2-carboxamidine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Trifluoromethyl)picolinimidamide hydrochloride
PC48656-1G
PC48656-250MG
APOSPC48656-1GEA
348
EUR
APOSPC48656-1G
APOSPC48656-250MG
4-(Trifluoromethyl)pyridine-2-carboxamidine hydrochloride
4-(Trifluoromethyl)pyridine-2-carboxamidine hydrochloride
Formel:
C₇H₆F₃N₃·ClH Molecular Weight: 225,599 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12755592 CAS-Nummer: 909109-68-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...