Methyl 3-amino-4-(trifluoromethyl)-1-benzothiophene-2-carboxylate

Lieferant: Apollo Scientific
Achtung

PC48667-1G PC48667-250MG
APOSPC48667-1GEA 208 EUR
APOSPC48667-1G APOSPC48667-250MG
Methyl 3-amino-4-(trifluoromethyl)-1-benzothiophene-2-carboxylate
Methyl 3-amino-4-(trifluoromethyl)-1-benzothiophene-2-carboxylate
Formel: C₁₁H₈F₃NO₂S
Molecular Weight: 275.247 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29169504
CAS-Nummer: 2379918-61-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR