3-Bromo-2,5-difluorobenzyl alcohol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Bromo-2,5-difluorophenyl)methanol

PC48752-250MG PC48752-1G PC48752-5G
APOSPC48752-250MGEA 69.6 EUR
APOSPC48752-250MG APOSPC48752-1G APOSPC48752-5G
3-Bromo-2,5-difluorobenzyl alcohol 98%
3-Bromo-2,5-difluorobenzyl alcohol
Formel: C₇H₅BrF₂O
Molecular Weight: 223.015 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27923603
CAS-Nummer: 1159186-56-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR