2-Amino-3-nitro-5-(trifluoromethyl)benzoic acid 95%

Lieferant: Apollo Scientific
Achtung

PC48760-1G PC48760-250MG
APOSPC48760-1GEA 719 EUR
APOSPC48760-1G APOSPC48760-250MG
2-Amino-3-nitro-5-(trifluoromethyl)benzoic acid 95%
2-Amino-3-nitro-5-(trifluoromethyl)benzoic acid
Formel: C₈H₅F₃N₂O₄
Molecular Weight: 250.132 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11847296
CAS-Nummer: 22227-60-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR