2,3,4-Trifluoro-6-(trifluoromethyl)benzyl alcohol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: [2,3,4-Trifluoro-6-(trifluoromethyl)phenyl]methanol

PC48777-1G PC48777-250MG PC48777-5G
APOSPC48777-1GEA 493 EUR
APOSPC48777-1G APOSPC48777-250MG APOSPC48777-5G
2,3,4-Trifluoro-6-(trifluoromethyl)benzyl alcohol 95%
2,3,4-Trifluoro-6-(trifluoromethyl)benzyl alcohol
Formel: C₈H₄F₆O
Molecular Weight: 230,107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31629891
CAS-Nummer: 2244085-32-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR