7-(Trifluoromethyl)quinazolin-4(3H)-one

Lieferant: Apollo Scientific
Achtung

PC48779-1G PC48779-250MG
APOSPC48779-1GEA 353 EUR
APOSPC48779-1G APOSPC48779-250MG
7-(Trifluoromethyl)quinazolin-4(3H)-one
7-(Trifluoromethyl)quinazolin-4(3H)-one
Formel: C₉H₅F₃N₂O
Molecular Weight: 214,144 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09954857
CAS-Nummer: 16499-58-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR