2,6-Dinitro-4-(trifluoromethyl)benzonitrile
Lieferant: Apollo Scientific
|
Achtung
|
PC48780-1G
PC48780-250MG
APOSPC48780-1GEA
161
EUR
APOSPC48780-1G
APOSPC48780-250MG
2,6-Dinitro-4-(trifluoromethyl)benzonitrile
2,6-Dinitro-4-(trifluoromethyl)benzonitrile
Formel:
C₈H₂F₃N₃O₄ Molecular Weight: 261.114 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD17168414 CAS-Nummer: 35213-02-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...