4-Methyl-6-(trifluoromethyl)pyrimidine-2-thiol
Lieferant: Apollo Scientific
|
Achtung
|
PC48781-1G
PC48781-250MG
APOSPC48781-1GEA
336
EUR
APOSPC48781-1G
APOSPC48781-250MG
4-Methyl-6-(trifluoromethyl)pyrimidine-2-thiol
4-Methyl-6-(trifluoromethyl)pyrimidine-2-thiol
Formel:
C₆H₅F₃N₂S Molecular Weight: 194,178 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01459879 CAS-Nummer: 78018-17-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...