Methyl 4-bromo-2,3,6-trifluorobenzoate 95%

Lieferant: Apollo Scientific

PC48810-1G PC48810-250MG PC48810-5G
APOSPC48810-1GEA 502 EUR
APOSPC48810-1G APOSPC48810-250MG APOSPC48810-5G
Methyl 4-bromo-2,3,6-trifluorobenzoate 95%
Methyl 4-bromo-2,3,6-trifluorobenzoate
Formel: C₈H₄BrF₃O₂
Molecular Weight: 269,015 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06204171
CAS-Nummer: 773134-13-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR