4-Bromo-2-chloro-5-fluorobenzotrifluoride 95%
Lieferant: Apollo Scientific
Synonyme:
1-Bromo-5-chloro-2-fluoro-4-(trifluoromethyl)benzene
PC48839-1G
PC48839-250MG
APOSPC48839-1GEA
540
EUR
APOSPC48839-1G
APOSPC48839-250MG
4-Bromo-2-chloro-5-fluorobenzotrifluoride 95%
4-Bromo-2-chloro-5-fluorobenzotrifluoride
Formel:
C₇H₂BrClF₄ Molecular Weight: 277,441 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD32869483 CAS-Nummer: 2169005-64-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...