Methyl 4-fluoro-3-sulphamoylbenzoate

Lieferant: Apollo Scientific
Achtung

PC48923-1G PC48923-250MG
APOSPC48923-1GEA 632 EUR
APOSPC48923-1G APOSPC48923-250MG
Methyl 4-fluoro-3-sulphamoylbenzoate
Methyl 4-fluoro-3-sulphamoylbenzoate
Formel: C₈H₈FNO₄S
Molecular Weight: 233,217 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11650724
CAS-Nummer: 85338-72-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR