(2,6-Difluoro-3-pyridin-3-yl)methanol

Lieferant: Apollo Scientific

Synonyme: (2,6-Difluoro-3-pyridyl)methanol

PC48932-1G PC48932-500MG PC48932-5G
APOSPC48932-1GEA 281 EUR
APOSPC48932-1G APOSPC48932-500MG APOSPC48932-5G
(2,6-Difluoro-3-pyridin-3-yl)methanol
(2,6-Difluoro-3-pyridin-3-yl)methanol
Formel: C₆H₅F₂NO
Molecular Weight: 145,107 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12911481
CAS-Nummer: 1105510-11-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR