4-(1,1,2,2-Tetrafluoroethoxy)benzeneboronic acid 95%
Lieferant: Apollo Scientific
Synonyme:
[4-(1,1,2,2-tetrafluoroethoxy)phenyl]boronic acid
PC48961-1G
PC48961-250MG
APOSPC48961-1GEA
574
EUR
APOSPC48961-1G
APOSPC48961-250MG
4-(1,1,2,2-Tetrafluoroethoxy)benzeneboronic acid 95%
4-(1,1,2,2-Tetrafluoroethoxy)benzeneboronic acid
Formel:
C₈H₇BF₄O₃ Molecular Weight: 237,944 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
1254118-45-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...