2-Bromo-3-fluoro-6-nitroanisole

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-1-fluoro-3-methoxy-4-nitrobenzene, 3-Bromo-4-fluoro-2-methoxynitrobenzene

PC49006-1G PC49006-250MG
APOSPC49006-1GEA 157 EUR
APOSPC49006-1G APOSPC49006-250MG
2-Bromo-3-fluoro-6-nitroanisole
2-Bromo-3-fluoro-6-nitroanisole
Formel: C₇H₅BrFNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18071062
CAS-Nummer: 1257535-01-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR