3-Bromo-4-fluoro-2-hydroxybenzonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-6-cyano-3-fluorophenol

PC49010-1G PC49010-250MG
APOSPC49010-1GEA 116 EUR
APOSPC49010-1G APOSPC49010-250MG
3-Bromo-4-fluoro-2-hydroxybenzonitrile
3-Bromo-4-fluoro-2-hydroxybenzonitrile
Formel: C₇H₃BrFNO
Lagertemperatur: Raumtemperatur
MDL: MFCD18071065
CAS-Nummer: 1257535-02-6
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR