3,3,3-Trifluoropropane-1,2-diol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Dihydroxy-1,1,1-trifluoropropane, 3,3,3-Trifluoropropylene glycol

PC49012-1G PC49012-5G PC49012-250MG
APOSPC49012-1GEA 102 EUR
APOSPC49012-1G APOSPC49012-5G APOSPC49012-250MG
3,3,3-Trifluoropropane-1,2-diol 97%
3,3,3-Trifluoropropane-1,2-diol
Formel: C₃H₅F₃O₂
Molecular Weight: 130,066 g/mol
Siedepunkt: 87…88 °C (25 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD02183521
CAS-Nummer: 431-39-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR