(Z)-1-Chloro-2-fluoroethylene 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
cis-1-Chloro-2-fluoroethylene, (Z)-1-Chloro-2-fluoroethene
PC49016-25G
PC49016-5G
APOSPC49016-25GEA
684
EUR
APOSPC49016-25G
APOSPC49016-5G
(Z)-1-Chloro-2-fluoroethylene 97%
(Z)-1-Chloro-2-fluoroethylene
Formel:
C₂H₂ClF Molecular Weight: 80,489 g/mol Siedepunkt: 15…15,5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08461574 CAS-Nummer: 2268-31-7 UN: 3161 ADR: 2.1, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...