3-Chloro-5-fluoropyridine-2-carbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Chloro-5-fluoropicolinonitrile, 3-Chloro-2-cyano-5-fluoropyridine
PC49019-100MG
PC49019-250MG
PC49019-1G
APOSPC49019-100MGEA
168
EUR
APOSPC49019-100MG
APOSPC49019-250MG
APOSPC49019-1G
3-Chloro-5-fluoropyridine-2-carbonitrile
3-Chloro-5-fluoropyridine-2-carbonitrile
Formel:
C₆H₂ClFN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD18255335 CAS-Nummer: 950670-25-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...