4-Bromo-2-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-α,α,α-trifluoro-2-picoline

PC49026-1G PC49026-5G PC49026-10G
APOSPC49026-1GEA 48 EUR
APOSPC49026-1G APOSPC49026-5G APOSPC49026-10G
4-Bromo-2-(trifluoromethyl)pyridine
4-Bromo-2-(trifluoromethyl)pyridine
Formel: C₆H₃BrF₃N
Molecular Weight: 225,994 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07774089
CAS-Nummer: 887583-90-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR