7-Fluoroquinoline-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Carboxy-7-fluoroquinoline

PC49032-1G PC49032-250MG
APOSPC49032-1GEA 491 EUR
APOSPC49032-1G APOSPC49032-250MG
7-Fluoroquinoline-3-carboxylic acid
7-Fluoroquinoline-3-carboxylic acid
Formel: C₁₀H₆FNO₂
Molecular Weight: 191.159 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787869
CAS-Nummer: 734524-15-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR