3-[4-Fluoro-2-(trifluoromethyl)phenyl]propanoic acid 98%
Lieferant: Apollo Scientific
Synonyme:
3-[4-Fluoro-2-(trifluoromethyl)phenyl]propionic acid, 4-Fluoro-2-(trifluoromethyl)hydrocinnamic acid
PC49072-1G
PC49072-5G
APOSPC49072-1GEA
87
EUR
APOSPC49072-1G
APOSPC49072-5G
3-[4-Fluoro-2-(trifluoromethyl)phenyl]propanoic acid 98%
3-[4-Fluoro-2-(trifluoromethyl)phenyl]propanoic acid
Formel:
C₁₀H₈F₄O₂ Molecular Weight: 236.163 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09025397 CAS-Nummer: 916420-73-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...