2'-Bromo-2,2,2,5'-tetrafluoroacetophenone

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Bromo-5-fluorophenyl)-2,2,2-trifluoroethan-1-one

PC49073-1G PC49073-250MG
APOSPC49073-1GEA 720 EUR
APOSPC49073-1G APOSPC49073-250MG
2'-Bromo-2,2,2,5'-tetrafluoroacetophenone
2'-Bromo-2,2,2,5'-tetrafluoroacetophenone
Formel: C₈H₃BrF₄O
Molecular Weight: 271,006 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25541883
CAS-Nummer: 1445995-82-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR