2-Fluoro-4-nitrobenzyl bromide 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
α-Bromo-2-fluoro-4-nitrotoluene, 1-(Bromomethyl)-2-fluoro-4-nitrobenzene
PC49076-1G
PC49076-250MG
PC49076-5G
APOSPC49076-1GEA
95.7
EUR
APOSPC49076-1G
APOSPC49076-250MG
APOSPC49076-5G
2-Fluoro-4-nitrobenzyl bromide 97%
2-Fluoro-4-nitrobenzyl bromide
Formel:
C₇H₅BrFNO₂ Molecular Weight: 234,023 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10000853 CAS-Nummer: 127349-56-8 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...