3-Amino-6-bromo-2-hydroxybenzotrifluoride 96%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 6-Amino-3-bromo-2-(trifluoromethyl)phenol, 4-Bromo-2-hydroxy-3-(trifluoromethyl)aniline

PC49077-1G PC49077-250MG
APOSPC49077-1GEA 406 EUR
APOSPC49077-1G APOSPC49077-250MG
3-Amino-6-bromo-2-hydroxybenzotrifluoride 96%
3-Amino-6-bromo-2-hydroxybenzotrifluoride
Formel: C₇H₅BrF₃NO
Molecular Weight: 256,02 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD25541884
CAS-Nummer: 1445995-79-8
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR