Ethyl 2-(3-{[4-(trifluoromethoxy)anilino]carbonyl}pyridin-2-yl)acetate 97%

Lieferant: Apollo Scientific

PC4908-1G PC4908-500MG
APOSPC4908-1GEA 977 EUR
APOSPC4908-1G APOSPC4908-500MG
Ethyl 2-(3-{[4-(trifluoromethoxy)anilino]carbonyl}pyridin-2-yl)acetate 97%
Ethyl 2-(3-{[4-(trifluoromethoxy)anilino]carbonyl}pyridin-2-yl)acetate
Formel: C₁₇H₁₅F₃N₂O₄
Molecular Weight: 368.307 g/mol
Schmelzpunkt: 105…107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00974993
CAS-Nummer: 338392-13-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR