4'-Bromo-2,2,2,2'-tetrafluoroacetophenone

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Bromo-2-fluorophenyl)-2,2,2-trifluoroethan-1-one, 1-(4-Bromo-2-fluorophenyl)-2,2,2-trifluoroethanone

PC49089-1G PC49089-250MG PC49089-5G
APOSPC49089-1GEA 150 EUR
APOSPC49089-1G APOSPC49089-250MG APOSPC49089-5G
4'-Bromo-2,2,2,2'-tetrafluoroacetophenone
4'-Bromo-2,2,2,2'-tetrafluoroacetophenone
Formel: C₈H₃BrF₄O
Molecular Weight: 271,006 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19439730
CAS-Nummer: 617706-18-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR