(Trifluoromethoxy)acetic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Trifluoromethoxy)ethanoic acid
PC49092-1G
PC49092-500MG
APOSPC49092-1GEA
415
EUR
APOSPC49092-1G
APOSPC49092-500MG
(Trifluoromethoxy)acetic acid
(Trifluoromethoxy)acetic acid
Formel:
C₃H₃F₃O₃ Siedepunkt: 84 °C (33 mm Hg) Dichte: 1,5014 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD11858185 CAS-Nummer: 69105-00-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...