3,5-Bis(trifluoromethyl)-α,N-dimethylbenzylamine

Lieferant: Apollo Scientific
Achtung

Synonyme: [1-[3,5-Bis(trifluoromethyl)phenyl]ethyl]methylamine, 1-[3,5-Bis(trifluoromethyl)phenyl]-N-methylethylamine

PC49094-1G PC49094-5G
APOSPC49094-1GEA 78.3 EUR
APOSPC49094-1G APOSPC49094-5G
3,5-Bis(trifluoromethyl)-α,N-dimethylbenzylamine
3,5-Bis(trifluoromethyl)-α,N-dimethylbenzylamine
Formel: C₁₁H₁₁F₆N
Molecular Weight: 271,202 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08460499
CAS-Nummer: 290297-43-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR