5-Bromo-6-chloro-2,2-difluoro-1,3-benzodioxole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-2-chloro-4,5-(difluoromethylenedioxy)benzene
PC49120-1G
PC49120-250MG
APOSPC49120-1GEA
717
EUR
APOSPC49120-1G
APOSPC49120-250MG
5-Bromo-6-chloro-2,2-difluoro-1,3-benzodioxole
5-Bromo-6-chloro-2,2-difluoro-1,3-benzodioxole
Formel:
C₇H₂BrClF₂O₂ Molecular Weight: 271,443 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09759045 CAS-Nummer: 869188-52-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...