(2,6-Difluorophenyl)methanesulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: α-(Chlorosulphonyl)-2,6-difluorotoluene

PC49124-1G PC49124-250MG
APOSPC49124-1GEA 429 EUR
APOSPC49124-1G APOSPC49124-250MG
(2,6-Difluorophenyl)methanesulphonyl chloride
(2,6-Difluorophenyl)methanesulphonyl chloride
Formel: C₇H₅ClF₂O₂S
Molecular Weight: 226.628 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11976096
CAS-Nummer: 179524-60-8
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR