2-Ethynyl-5-fluoro-3-methylpyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: (5-Fluoro-3-methylpyridin-2-yl)acetylene, 2-Ethynyl-5-fluoro-3-picoline

PC49135-1G PC49135-250MG
APOSPC49135-1GEA 650 EUR
APOSPC49135-1G APOSPC49135-250MG
2-Ethynyl-5-fluoro-3-methylpyridine
2-Ethynyl-5-fluoro-3-methylpyridine
Formel: C₈H₆FN
Molecular Weight: 135.138 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD21648440
CAS-Nummer: 1372103-93-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR