2-Ethynyl-3-fluoro-5-methylpyridine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Ethynyl-5-fluoro-3-picoline, (3-Fluoro-5-methylpyridin-2-yl)acetylene
PC49136-1G
PC49136-250MG
APOSPC49136-1GEA
154
EUR
APOSPC49136-1G
APOSPC49136-250MG
2-Ethynyl-3-fluoro-5-methylpyridine
2-Ethynyl-3-fluoro-5-methylpyridine
Formel:
C₈H₆FN Molecular Weight: 135.138 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD21648441 CAS-Nummer: 1372103-91-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...