3,3,3-Trifluoropropanal hydrate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,3,3-Trifluoropropionaldehyde hydrate, 3,3,3-Trifluoropropaldehyde hydrate, 3,3,3-Trifluoropropionaldehyde hydrate (diol)
PC49156-25G
PC49156-5G
APOSPC49156-25GEA
865
EUR
APOSPC49156-25G
APOSPC49156-5G
3,3,3-Trifluoropropanal hydrate
3,3,3-Trifluoropropanal hydrate
Formel:
C₃H₃F₃O·H₂O Molecular Weight: 130,066 g/mol Siedepunkt: 56…56,5 °C (745 mm Hg) Dichte: 1,3009 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD18384868 CAS-Nummer: 1309602-82-1 UN: 1989 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...