2'-Amino-2,2,2-trifluoro-5'-(trifluoromethyl)acetophenone

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-[2-Amino-5-(trifluoromethyl)phenyl]-2,2,2-trifluoroethan-1-one, 2-(Trifluoroacetyl)-4-(trifluoromethyl)aniline, 4-Amino-3-(trifluoroacetyl)benzotrifluoride, 1-(2-Amino-5-(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone

PC49165-1G PC49165-250MG
APOSPC49165-1GEA 522 EUR
APOSPC49165-1G APOSPC49165-250MG
2'-Amino-2,2,2-trifluoro-5'-(trifluoromethyl)acetophenone
2'-Amino-2,2,2-trifluoro-5'-(trifluoromethyl)acetophenone
Formel: C₉H₅F₆NO
Lagertemperatur: Kühlschrank
MDL: MFCD18071074
CAS-Nummer: 489429-73-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR