3-Bromo-4-(trifluoromethoxy)benzoyl chloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Bromo-4-(chlorocarbonyl)-α,α,α-trifluoroanisole

PC49187-1G PC49187-5G
APOSPC49187-1GEA 231 EUR
APOSPC49187-1G APOSPC49187-5G
3-Bromo-4-(trifluoromethoxy)benzoyl chloride
3-Bromo-4-(trifluoromethoxy)benzoyl chloride
Formel: C₈H₃BrClF₃O₂
Molecular Weight: 303.46 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19441789
CAS-Nummer: 85366-63-8
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR