4-Fluoroindoline

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Dihydro-4-fluoro-1H-indole

PC49192-1G PC49192-250MG
APOSPC49192-1GEA 177 EUR
APOSPC49192-1G APOSPC49192-250MG
4-Fluoroindoline
4-Fluoroindoline
Formel: C₈H₈FN
Molecular Weight: 137.154 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07371634
CAS-Nummer: 552866-98-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR