3-(Difluoromethyl)benzyl alcohol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: [3-(Difluoromethyl)phenyl]methanol, 3-(Hydroxymethyl)benzal fluoride

PC49199-5G PC49199-1G
APOSPC49199-5GEA 1250 EUR
APOSPC49199-5G APOSPC49199-1G
3-(Difluoromethyl)benzyl alcohol 97%
3-(Difluoromethyl)benzyl alcohol
Formel: C₈H₈F₂O
Molecular Weight: 158.145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19053257
CAS-Nummer: 1309875-03-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR