4-Bromo-3-(trifluoromethoxy)benzonitrile
Lieferant: Apollo Scientific
|
Achtung
|
PC49242-1G
PC49242-250MG
PC49242-5G
APOSPC49242-1GEA
29
EUR
APOSPC49242-1G
APOSPC49242-250MG
APOSPC49242-5G
4-Bromo-3-(trifluoromethoxy)benzonitrile
4-Bromo-3-(trifluoromethoxy)benzonitrile
Formel:
C₈H₃BrF₃NO Molecular Weight: 266.015 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22378817 CAS-Nummer: 928136-78-1 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...