2-Bromo-5-(difluoromethyl)thiophene
Lieferant: Apollo Scientific
|
Achtung
|
PC49250-250MG
PC49250-500MG
APOSPC49250-250MGEA
42.2
EUR
APOSPC49250-250MG
APOSPC49250-500MG
2-Bromo-5-(difluoromethyl)thiophene
2-Bromo-5-(difluoromethyl)thiophene
Formel:
C₅H₃BrF₂S Molecular Weight: 213,043 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD22378823 CAS-Nummer: 924646-90-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...