5-Bromo-3-fluoro-N-methyl-2-nitroaniline
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(5-Bromo-3-fluoro-2-nitrophenyl)methylamine, 4-Bromo-2-fluoro-6-(methylamino)nitrobenzene
PC49258-1G
PC49258-250MG
APOSPC49258-1GEA
261
EUR
APOSPC49258-1G
APOSPC49258-250MG
5-Bromo-3-fluoro-N-methyl-2-nitroaniline
5-Bromo-3-fluoro-N-methyl-2-nitroaniline
Formel:
C₇H₆BrFN₂O₂ Molecular Weight: 249.037 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD22415277 CAS-Nummer: 1396503-81-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...